The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321270.1 | NP_001308199.1 | 2 - 3 | 297 | 70 |
XM_011532863.2 | XP_011531165.1 | 2 - 3 | 257 | 70 | |
XM_011532864.2 | XP_011531166.1 | 2 - 3 | 294 | 70 | |
XM_011532865.1 | XP_011531167.1 | 2 - 3 | 198 | 70 | |
GenBank mRNA | AK092257.1 | - | 2 - 3 | 198 | 70 |
DC372029.1 | - | 2 - 3 | 294 | 70 |