Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPN14
Entrez Gene ID
440854
Gene Name
calpain 14
Gene Aliases
-
Chromosome Location
Chr.2: 31173056 - 31233957 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321270.1 NP_001308199.1 5 - 6 757 74
XM_011532866.2 XP_011531168.1 4 - 5 562 74
GenBank mRNA AK092257.1 - 5 - 6 658 74

View Full Product Details