Product Details

Assay Reference Genome
Location

Chr.15:68207666 on build GRCh38
Cytoband
15q23
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
54982
Gene Symbol
CLN6
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Gene Aliases
CLN4A, HsT18960, nclf
Location
Chr.15:68206992-68229742 on build GRCh38
Assay Gene Location
Within Exon 7
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CLN6 NM_017882.2 7 1568 NP_060352.1
AK000568.1 7 1513 BAA91260.1
AK027604.1 7 1536 BAB55226.1
AK074753.1 4 1062 BAC11181.1
AK291175.1 7 1495
BC010849.1 7 1482 AAH10849.1
BC013130.1 7 1419 AAH13130.1
CA446197.1
CN427564.1 1 334

View Full Product Details