The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289020.1 | NP_001275949.1 | 1 - 2 | 157 | 107 |
NM_001289021.1 | NP_001275950.1 | 1 - 2 | 157 | 107 | |
NM_145027.4 | NP_659464.3 | 1 - 2 | 157 | 107 | |
XM_005248904.4 | XP_005248961.1 | 1 - 2 | 157 | 107 | |
XM_011514357.2 | XP_011512659.1 | 1 - 2 | 82 | 107 | |
XM_011514358.2 | XP_011512660.1 | 1 - 2 | 82 | 107 | |
XM_011514359.2 | XP_011512661.1 | 1 - 2 | 82 | 107 | |
XM_011514361.2 | XP_011512663.1 | 1 - 2 | 157 | 107 | |
XM_017010427.1 | XP_016865916.1 | 1 - 2 | 157 | 107 | |
GenBank mRNA | AK131471.1 | - | 1 - 2 | 157 | 107 |
BC117307.1 | - | 1 - 2 | 87 | 107 | |
BC143444.1 | - | 1 - 2 | 87 | 107 | |
BC143445.1 | - | 1 - 2 | 87 | 107 | |
DB080579.1 | - | 1 - 2 | 157 | 107 |