The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080529.2 | NP_001073998.2 | 6 - 7 | 1433 | 154 |
XM_017012522.1 | XP_016868011.1 | 5 - 6 | 1441 | 154 | |
GenBank mRNA | AK094250.1 | - | 2 - 3 | 590 | 154 |
BX107905.1 | - | 2 - 3 | 270 | 154 |