The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080464.2 | NP_001073933.2 | 1 - 2 | 169 | 83 |
XM_005267590.4 | XP_005267647.1 | 1 - 2 | 195 | 83 | |
XM_017021268.1 | XP_016876757.1 | 1 - 2 | 197 | 83 | |
GenBank mRNA | AX781433.1 | - | 1 - 2 | 176 | 83 |
AX781435.1 | - | 1 - 2 | 84 | 83 | |
BC035836.1 | - | 1 - 2 | 144 | 83 | |
BC136637.1 | - | 1 - 2 | 111 | 83 | |
BX247999.1 | - | 1 - 2 | 169 | 83 | |
BX280385.1 | - | 1 - 2 | 144 | 83 | |
BX407951.2 | - | 1 - 2 | 168 | 82 |