Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LINC00619
Entrez Gene ID
414260
Gene Name
long intergenic non-protein coding RNA 619
Gene Aliases
C10orf136, bA168P8.1
Chromosome Location
Chr.10: 43845306 - 43850622 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033923.1 1 - 2 96 70
GenBank mRNA BC017939.1 - 1 - 2 96 70


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BF670044.1 - 1 - 2 97 Chr. 10 - 43845306 - 43850199 [+] on Build GRCh38 70


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream heterogeneous nuclear ribonucleoprotein A3 pseudogene 1 HNRNPA3P1 D10S102, FBRNP, HNRPA3, HNRPA3P1 -54.9kb Anti-sense Chr. - 43787412 - 43790417 on Build GRCh38
Downstream long intergenic non-protein coding RNA 840 LINC00840 13.8kb Sense Chr. - 43859411 - 43895433 on Build GRCh38

View Full Product Details