The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032199.2 | NP_115575.1 | 2 - 3 | 732 | 60 |
XM_011540262.1 | XP_011538564.1 | 2 - 3 | 732 | 60 | |
GenBank mRNA | AK296921.1 | - | 2 - 3 | 298 | 60 |
BC107800.1 | - | 2 - 3 | 291 | 60 |