The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244638.1 | NP_001231567.1 | 2 - 3 | 385 | 82 |
NM_032199.2 | NP_115575.1 | 5 - 6 | 1301 | 82 | |
XM_011540262.1 | XP_011538564.1 | 4 - 5 | 1070 | 82 | |
XM_017016765.1 | XP_016872254.1 | 1 - 2 | 112 | 82 | |
GenBank mRNA | AK024803.1 | - | 2 - 3 | 130 | 82 |
BC015120.1 | - | 2 - 3 | 389 | 82 | |
BC066345.1 | - | 2 - 3 | 354 | 82 | |
BC107800.1 | - | 5 - 6 | 860 | 82 |