The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244638.1 | NP_001231567.1 | 6 - 7 | 935 | 140 |
NM_032199.2 | NP_115575.1 | 9 - 10 | 1851 | 140 | |
XM_011540262.1 | XP_011538564.1 | 8 - 9 | 1620 | 140 | |
XM_017016765.1 | XP_016872254.1 | 4 - 5 | 609 | 140 | |
GenBank mRNA | M73837.1 | - | 4 - 5 | 492 | 140 |