Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERMARD
Entrez Gene ID
55780
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C6orf70, PVNH6, dJ266L20.3
Chromosome Location
Chr.6: 169751622 - 169781584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278531.1 NP_001265460.1 16 - 17 1627 92
GenBank mRNA AK001369.1 - 15 - 16 1488 92

View Full Product Details