Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERMARD
Entrez Gene ID
55780
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C6orf70, PVNH6, dJ266L20.3
Chromosome Location
Chr.6: 169751622 - 169781584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278531.1 NP_001265460.1 7 - 8 777 150
NM_001278532.1 NP_001265461.1 6 - 7 608 150
NM_001278533.1 NP_001265462.1 7 - 8 777 150
NM_018341.2 NP_060811.1 7 - 8 777 150
XM_011535938.2 XP_011534240.1 7 - 8 777 150
XM_011535939.2 XP_011534241.1 7 - 8 1867 150
XM_011535940.2 XP_011534242.1 7 - 8 1676 150
XM_017011027.1 XP_016866516.1 7 - 8 1617 150
XM_017011028.1 XP_016866517.1 7 - 8 942 150
XM_017011029.1 XP_016866518.1 7 - 8 702 150
XM_017011030.1 XP_016866519.1 6 - 7 526 150
XM_017011031.1 XP_016866520.1 6 - 7 591 150
XM_017011032.1 XP_016866521.1 7 - 8 1862 150
XM_017011033.1 XP_016866522.1 7 - 8 1671 150
GenBank mRNA AK001369.1 - 6 - 7 638 150
AK294090.1 - 6 - 7 605 150
BC044812.1 - 7 - 8 761 150
CA396814.1 - 5 - 6 457 150

View Full Product Details