Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERMARD
Entrez Gene ID
55780
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C6orf70, PVNH6, dJ266L20.3
Chromosome Location
Chr.6: 169751622 - 169781584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278531.1 NP_001265460.1 9 - 10 994 129
NM_001278532.1 NP_001265461.1 8 - 9 825 129
NM_001278533.1 NP_001265462.1 9 - 10 994 129
NM_018341.2 NP_060811.1 9 - 10 994 129
XM_011535939.2 XP_011534241.1 9 - 10 2084 129
XM_011535940.2 XP_011534242.1 9 - 10 1893 129
XM_017011027.1 XP_016866516.1 9 - 10 1834 129
XM_017011028.1 XP_016866517.1 9 - 10 1159 129
XM_017011029.1 XP_016866518.1 9 - 10 919 129
XM_017011030.1 XP_016866519.1 8 - 9 743 129
XM_017011031.1 XP_016866520.1 8 - 9 808 129
XM_017011032.1 XP_016866521.1 9 - 10 2079 129
XM_017011033.1 XP_016866522.1 9 - 10 1888 129
GenBank mRNA AK001369.1 - 8 - 9 855 129
AK002014.1 - 9 - 10 991 129
AK294090.1 - 8 - 9 822 129
BC044812.1 - 9 - 10 978 129

View Full Product Details