Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM71E2
Entrez Gene ID
284418
Gene Name
family with sequence similarity 71 member E2
Gene Aliases
C19orf16
Chromosome Location
Chr.19: 55354908 - 55363252 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145402.1 NP_001138874.1 10 - 11 2994 75
GenBank mRNA AK127979.1 - 9 - 10 2975 75
AL834316.1 - 3 - 4 1752 75

View Full Product Details