Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CFAP69
- Entrez Gene ID
-
79846
- Gene Name
- cilia and flagella associated protein 69
- Gene Aliases
-
C7orf63, FAP69
- Chromosome Location
-
Chr.7: 90245174 - 90320964 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL137642.1 |
- |
1 - 2 |
322 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
STEAP2 metalloreductase |
STEAP2 |
IPCA1, PCANAP1, PUMPCn, STAMP1, STMP |
-74.8kb
|
Sense |
Chr. - 90211686 - 90243408 on Build GRCh38 |
Downstream |
uncharacterized LOC101927446 |
LOC101927446 |
|
14.8kb
|
Sense |
Chr. - 90335223 - 90344094 on Build GRCh38 |
View Full Product Details