Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC24A5
Entrez Gene ID
283652
Gene Name
solute carrier family 24 member 5
Gene Aliases
JSX, NCKX5, OCA6, SHEP4
Chromosome Location
Chr.15: 48120972 - 48142392 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205850.2 NP_995322.1 5 - 6 667 99
XM_017022079.1 XP_016877568.1 4 - 5 1768 99
XM_017022080.1 XP_016877569.1 4 - 5 1988 99
GenBank mRNA AF348468.1 - 5 - 6 667 99
BC113628.1 - 5 - 6 622 99
BC113630.1 - 5 - 6 622 99
BC143950.1 - 5 - 6 622 99
DQ665306.1 - 5 - 6 615 99
DQ665307.1 - 4 - 5 435 99

View Full Product Details