Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERMARD
Entrez Gene ID
55780
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C6orf70, PVNH6, dJ266L20.3
Chromosome Location
Chr.6: 169751622 - 169781584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278532.1 NP_001265461.1 14 - 15 1389 71
NM_018341.2 NP_060811.1 15 - 16 1558 71
XM_011535938.2 XP_011534240.1 14 - 15 1528 71
XM_011535939.2 XP_011534241.1 15 - 16 2648 71
XM_011535940.2 XP_011534242.1 15 - 16 2457 71
XM_017011027.1 XP_016866516.1 15 - 16 2398 71
XM_017011028.1 XP_016866517.1 15 - 16 1723 71
XM_017011029.1 XP_016866518.1 15 - 16 1483 71
XM_017011030.1 XP_016866519.1 14 - 15 1307 71
XM_017011031.1 XP_016866520.1 14 - 15 1372 71
XM_017011032.1 XP_016866521.1 15 - 16 2643 71
XM_017011033.1 XP_016866522.1 15 - 16 2452 71
GenBank mRNA AK002014.1 - 15 - 16 1555 71
AK021520.1 - 5 - 6 1622 71
AK294090.1 - 14 - 15 1386 71

View Full Product Details