The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013742.3 | NP_001013764.1 | 24 - 25 | 3468 | 78 |
GenBank mRNA | AB183864.1 | - | 24 - 25 | 3468 | 78 |
BC137319.1 | - | 24 - 25 | 3452 | 78 | |
BC137320.1 | - | 24 - 25 | 3452 | 78 | |
DB728104.1 | - | 2 - 3 | 242 | 78 |