Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYCE1L
Entrez Gene ID
100130958
Gene Name
synaptonemal complex central element protein 1 like
Gene Aliases
MRP2
Chromosome Location
Chr.16: 77199452 - 77213079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129979.1 NP_001123451.1 5 - 6 303 85

View Full Product Details