Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TTTY7
- Entrez Gene ID
-
246122
- Gene Name
- testis-specific transcript, Y-linked 7 (non-protein coding)
- Gene Aliases
-
CLONE795723, LINC00129, NCRNA00129, TTTY7A, TTTY7B, TTY7
- Chromosome Location
-
Chr.Y: 6449468 - 6457906 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001534.2 |
|
6 - 7 |
1117 |
73 |
View Full Product Details