The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306084.1 | NP_001293013.1 | 42 - 43 | 6048 | 94 |
XM_011539069.2 | XP_011537371.1 | 43 - 44 | 6248 | 94 | |
XM_011539070.2 | XP_011537372.1 | 43 - 44 | 6248 | 94 | |
XM_011539071.2 | XP_011537373.1 | 43 - 44 | 6248 | 94 | |
XM_011539072.2 | XP_011537374.1 | 43 - 44 | 6248 | 94 | |
XM_011539073.2 | XP_011537375.1 | 43 - 44 | 6248 | 94 | |
XM_011539074.2 | XP_011537376.1 | 43 - 44 | 6248 | 94 | |
XM_011539076.2 | XP_011537378.1 | 43 - 44 | 6248 | 94 | |
XM_011539077.2 | XP_011537379.1 | 42 - 43 | 6398 | 94 | |
XM_011539078.2 | XP_011537380.1 | 43 - 44 | 6248 | 94 | |
XM_011539079.1 | XP_011537381.1 | 39 - 40 | 5494 | 94 | |
XM_011539081.2 | XP_011537383.1 | 43 - 44 | 6248 | 94 | |
XM_011539083.2 | XP_011537385.1 | 43 - 44 | 6248 | 94 | |
XM_011539084.2 | XP_011537386.1 | 43 - 44 | 6248 | 94 | |
XM_011539085.1 | XP_011537387.1 | 28 - 29 | 4069 | 94 | |
XM_017018866.1 | XP_016874355.1 | 43 - 44 | 6248 | 94 | |
XM_017018867.1 | XP_016874356.1 | 32 - 33 | 4500 | 94 | |
GenBank mRNA | AK126100.1 | - | 9 - 10 | 1362 | 94 |