Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTTY22
Entrez Gene ID
252954
Gene Name
testis-specific transcript, Y-linked 22 (non-protein coding)
Gene Aliases
NCRNA00147
Chromosome Location
Chr.Y: 9801153 - 9813245 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_001539.1 4 - 5 377 92


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF527834.3 - 4 - 5 377 Chr. Y - 9801153 - 9813245 [+] on Build GRCh38 92


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream testis-specific transcript, Y-linked 1 (non-protein coding) TTTY1 LINC00033, NCRNA00033, TTY1 -35.8kb Anti-sense Chr. - 9753156 - 9774319 on Build GRCh38
Downstream testis-specific transcript, Y-linked 23 (non-protein coding) TTTY23 NCRNA00148, TTTY23B 97.6kb Sense Chr. - 9910798 - 9911962 on Build GRCh38

View Full Product Details