The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042367.1 | NP_001035826.1 | 1 - 2 | 187 | 135 |
GenBank mRNA | AK131030.1 | - | 1 - 2 | 181 | 135 |
BC055412.1 | - | 1 - 2 | 168 | 135 |