The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267723.1 | NP_001254652.1 | 4 - 5 | 440 | 82 |
XM_005259191.3 | XP_005259248.1 | 4 - 5 | 2545 | 82 | |
XM_005259192.4 | XP_005259249.1 | 4 - 5 | 1186 | 82 | |
XM_011527257.2 | XP_011525559.1 | 4 - 5 | 469 | 82 | |
GenBank mRNA | DR007040.1 | - | 4 - 5 | 460 | 82 |
GQ375050.1 | - | 4 - 5 | 413 | 82 |