Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC61
Entrez Gene ID
729440
Gene Name
coiled-coil domain containing 61
Gene Aliases
-
Chromosome Location
Chr.19: 45995457 - 46018616 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267723.1 NP_001254652.1 6 - 7 819 51
XM_005259191.3 XP_005259248.1 6 - 7 2924 51
XM_005259192.4 XP_005259249.1 6 - 7 1565 51
XM_011527257.2 XP_011525559.1 6 - 7 848 51
GenBank mRNA AW453013.1 - 2 - 3 51

View Full Product Details