The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005335.1 | NP_001005335.1 | 3 - 4 | 403 | 66 |
NM_001533.2 | NP_001524.2 | 3 - 4 | 637 | 66 | |
XM_011526886.1 | XP_011525188.1 | 3 - 4 | 426 | 66 | |
XM_011526887.1 | XP_011525189.1 | 3 - 4 | 470 | 66 | |
XM_011526889.1 | XP_011525191.1 | 3 - 4 | 521 | 66 | |
GenBank mRNA | AB044547.1 | - | 3 - 4 | 632 | 66 |
AK292115.1 | - | 3 - 4 | 402 | 66 | |
AK298384.1 | - | 3 - 4 | 473 | 66 | |
AK301062.1 | - | 3 - 4 | 411 | 66 | |
AK313652.1 | - | 3 - 4 | 572 | 66 | |
BC069184.1 | - | 3 - 4 | 576 | 66 | |
BI256815.1 | - | 3 - 4 | 403 | 66 | |
X16135.1 | - | 3 - 4 | 561 | 66 |