The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005335.1 | NP_001005335.1 | 9 - 10 | 1135 | 86 |
NM_001533.2 | NP_001524.2 | 9 - 10 | 1369 | 86 | |
XM_011526886.1 | XP_011525188.1 | 9 - 10 | 1158 | 86 | |
XM_011526887.1 | XP_011525189.1 | 9 - 10 | 1202 | 86 | |
XM_011526888.2 | XP_011525190.1 | 7 - 8 | 1257 | 86 | |
XM_011526889.1 | XP_011525191.1 | 9 - 10 | 1253 | 86 | |
GenBank mRNA | AB044547.1 | - | 9 - 10 | 1364 | 86 |
AK092828.1 | - | 3 - 4 | 1668 | 86 | |
AK094966.1 | - | 2 - 3 | 2359 | 86 | |
AK097975.1 | - | 1 - 2 | 774 | 86 | |
AK292115.1 | - | 9 - 10 | 1134 | 86 | |
AK313652.1 | - | 9 - 10 | 1304 | 86 | |
BC069184.1 | - | 9 - 10 | 1308 | 86 | |
X16135.1 | - | 9 - 10 | 1293 | 86 |