The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243476.2 | NP_001230405.1 | 3 - 4 | 473 | 135 |
XM_011535299.2 | XP_011533601.1 | 2 - 3 | 219 | 135 | |
GenBank mRNA | BX647695.1 | - | 3 - 4 | 433 | 135 |
DA312208.1 | - | 2 - 3 | 163 | 135 |