Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DLGAP1-AS5
- Entrez Gene ID
-
284215
- Gene Name
- DLGAP1 antisense RNA 5
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 4264602 - 4296000 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_036489.1 |
|
2 - 3 |
383 |
139 |
GenBank mRNA |
AK098180.1 |
- |
2 - 3 |
352 |
139 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB205872.1 |
- |
2 - 3 |
352 |
|
Chr. 18 - 4264633 - 4293412 [+] on Build GRCh38 |
139 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
DLGAP1 antisense RNA 4 |
DLGAP1-AS4 |
|
-261.4kb
|
Sense |
Chr. - 3962353 - 4013943 on Build GRCh38 |
Downstream |
A kinase (PRKA) anchor inhibitor 1 |
AKAIN1 |
C18orf42 |
848.3kb
|
Anti-sense |
Chr. - 5143673 - 5199997 on Build GRCh38 |
View Full Product Details