The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020225.2 | NP_064610.1 | 3 - 4 | 4017 | 88 |
XM_011532129.1 | XP_011530431.1 | 3 - 4 | 4141 | 88 | |
XM_011532131.1 | XP_011530433.1 | 6 - 7 | 3272 | 88 | |
XM_017008467.1 | XP_016863956.1 | 4 - 5 | 3178 | 88 | |
GenBank mRNA | AB037813.1 | - | 3 - 4 | 4017 | 88 |
AK289840.1 | - | 3 - 4 | 2696 | 88 | |
BC146754.1 | - | 3 - 4 | 4017 | 88 |