The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101341.1 | NP_001094811.1 | 1 - 2 | 671 | 64 |
GenBank mRNA | BC042093.1 | - | 1 - 2 | 671 | 64 |
BC129834.1 | - | 1 - 2 | 417 | 64 | |
KF856952.1 | - | 1 - 2 | 766 | 64 |