The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204118.1 | NP_001191047.1 | 10 - 11 | 723 | 122 |
NM_207390.3 | NP_997273.3 | 10 - 11 | 723 | 122 | |
NR_109785.1 | 10 - 11 | 723 | 122 | ||
XM_017026785.1 | XP_016882274.1 | 11 - 12 | 1267 | 122 | |
XM_017026786.1 | XP_016882275.1 | 11 - 12 | 1131 | 122 | |
XM_017026787.1 | XP_016882276.1 | 12 - 13 | 1486 | 122 | |
XM_017026788.1 | XP_016882277.1 | 11 - 12 | 1243 | 122 | |
XM_017026789.1 | XP_016882278.1 | 7 - 8 | 945 | 122 | |
XM_017026790.1 | XP_016882279.1 | 10 - 11 | 1219 | 122 | |
XM_017026792.1 | XP_016882281.1 | 11 - 12 | 1271 | 122 | |
XM_017026793.1 | XP_016882282.1 | 11 - 12 | 1274 | 122 | |
GenBank mRNA | BC140848.1 | - | 10 - 11 | 684 | 122 |
BC144665.1 | - | 10 - 11 | 684 | 122 |