Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUTM1
Entrez Gene ID
256646
Gene Name
NUT midline carcinoma family member 1
Gene Aliases
C15orf55, FAM22H, NUT
Chromosome Location
Chr.15: 34343315 - 34357737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284292.1 NP_001271221.1 5 - 6 1459 59
NM_001284293.1 NP_001271222.1 4 - 5 1365 59
NM_175741.2 NP_786883.1 5 - 6 1544 59
GenBank mRNA AF482429.1 - 4 - 5 1148 59
AK302656.1 - 5 - 6 1459 59
BC114518.1 - 4 - 5 1120 59
BC114567.1 - 4 - 5 1115 59

View Full Product Details