Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH12
Entrez Gene ID
201625
Gene Name
dynein axonemal heavy chain 12
Gene Aliases
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
Chromosome Location
Chr.3: 57293699 - 57545203 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291661.1 NP_001278590.1 22 - 23 3522 77
XM_011533466.1 XP_011531768.1 22 - 23 3519 77
XM_011533467.1 XP_011531769.1 22 - 23 3486 77
XM_011533468.1 XP_011531770.1 22 - 23 3519 77
XM_011533469.1 XP_011531771.1 22 - 23 3519 77
XM_011533471.2 XP_011531773.1 22 - 23 3519 77
XM_017005860.1 XP_016861349.1 22 - 23 3519 77
XM_017005861.1 XP_016861350.1 21 - 22 4202 77

View Full Product Details