Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GADL1
Entrez Gene ID
339896
Gene Name
glutamate decarboxylase like 1
Gene Aliases
ADC, CSADC, HuADC, HuCSADC
Chromosome Location
Chr.3: 30726009 - 30894661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207359.2 NP_997242.2 7 - 8 775 71
XM_017006297.1 XP_016861786.1 7 - 8 1173 71
GenBank mRNA AK128643.1 - 2 - 3 204 71
AL832766.1 - 7 - 8 775 71
BC093701.1 - 2 - 3 198 71
BC111986.1 - 2 - 3 198 71

View Full Product Details