Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCR4
Entrez Gene ID
1233
Gene Name
C-C motif chemokine receptor 4
Gene Aliases
CC-CKR-4, CD194, CKR4, CMKBR4, ChemR13, HGCN:14099, K5-5
Chromosome Location
Chr.3: 32951555 - 32955312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017005687.1 XP_016861176.1 1 - 2 136 74

View Full Product Details