The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213600.3 | NP_998765.3 | 7 - 8 | 696 | 121 |
NR_033151.1 | 6 - 7 | 623 | 121 | ||
GenBank mRNA | AK122623.1 | - | 7 - 8 | 667 | 121 |
AK304368.1 | - | 7 - 8 | 678 | 121 | |
BC035335.1 | - | 6 - 7 | 593 | 121 | |
BC142682.1 | - | 7 - 8 | 667 | 121 | |
BC146648.1 | - | 7 - 8 | 667 | 121 |