The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207444.2 | NP_997327.1 | 1 - 2 | 136 | 81 |
GenBank mRNA | AK093014.1 | - | 1 - 2 | 136 | 81 |
BC119016.1 | - | 1 - 2 | 136 | 81 | |
BC119810.1 | - | 1 - 2 | 136 | 81 |