Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NAALADL2
- Entrez Gene ID
-
254827
- Gene Name
- N-acetylated alpha-linked acidic dipeptidase-like 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 174439669 - 175809510 on Build GRCh38
View Full Product Details