Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFMBT2
Entrez Gene ID
57713
Gene Name
Scm-like with four mbt domains 2
Gene Aliases
-
Chromosome Location
Chr.10: 7158624 - 7411486 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018039.1 NP_001018049.1 14 - 15 1651 147
NM_001029880.2 NP_001025051.1 14 - 15 1753 147
XM_005252551.2 XP_005252608.1 14 - 15 1534 147
XM_006717490.2 XP_006717553.1 12 - 13 1526 147
XM_011519611.1 XP_011517913.1 14 - 15 1752 147
XM_017016466.1 XP_016871955.1 14 - 15 1654 147
GenBank mRNA AB046837.1 - 14 - 15 1651 147
BC152430.1 - 14 - 15 1651 147

View Full Product Details