The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012309.4 | NP_036441.2 | 3 - 4 | 521 | 85 |
XM_017017387.1 | XP_016872876.1 | 4 - 5 | 626 | 85 | |
XM_017017388.1 | XP_016872877.1 | 4 - 5 | 648 | 85 | |
XM_017017389.1 | XP_016872878.1 | 4 - 5 | 821 | 85 | |
GenBank mRNA | AB208025.1 | - | 3 - 4 | 487 | 85 |
DQ152234.1 | - | 2 - 3 | 409 | 85 |