Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HNRNPUL2
- Entrez Gene ID
-
221092
- Gene Name
- heterogeneous nuclear ribonucleoprotein U like 2
- Gene Aliases
-
HNRPUL2, SAF-A2
- Chromosome Location
-
Chr.11: 62712625 - 62727385 on Build GRCh38
- Gene Symbol
- HNRNPUL2-BSCL2
- Entrez Gene ID
-
100534595
- Gene Name
- HNRNPUL2-BSCL2 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 62690262 - 62727384 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037946.1 |
|
12 - 13 |
2277 |
98 |
View Full Product Details