Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC10A6
Entrez Gene ID
345274
Gene Name
solute carrier family 10 member 6
Gene Aliases
SOAT
Chromosome Location
Chr.4: 86823468 - 86849263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_197965.2 NP_932069.1 3 - 4 733 102
GenBank mRNA AJ583502.1 - 3 - 4 585 102
BC107051.1 - 3 - 4 585 102
BC107052.1 - 3 - 4 585 102
EF437223.1 - 3 - 4 733 102

View Full Product Details