The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_197965.2 | NP_932069.1 | 4 - 5 | 909 | 129 |
GenBank mRNA | AJ583502.1 | - | 4 - 5 | 761 | 129 |
BC107051.1 | - | 4 - 5 | 761 | 129 | |
BC107052.1 | - | 4 - 5 | 761 | 129 | |
EF437223.1 | - | 4 - 5 | 909 | 129 |