Product Details
- Assay Reference Genome
Location
Chr.9:108899836 on build GRCh38
- Cytoband
- 9q31.3
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 8518
- Gene Symbol
- IKBKAP
- Gene Name
- inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
- Gene Aliases
- DYS, ELP1, FD, IKAP, IKI3, TOT1
- Location
- Chr.9:108867520-108934332 on build GRCh38
- Assay Gene Location
- Overlaps Intron 20 - Exon 20
View Full Product Details