Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDC
Entrez Gene ID
10726
Gene Name
nuclear distribution C, dynein complex regulator
Gene Aliases
HNUDC, MNUDC, NPD011
Chromosome Location
Chr.1: 26911128 - 26946871 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006600.3 NP_006591.1 7 - 8 955 76
XM_011540529.1 XP_011538831.1 7 - 8 855 76
XM_011540530.1 XP_011538832.1 7 - 8 1093 76
XM_017000094.1 XP_016855583.1 8 - 9 1026 76
XM_017000095.1 XP_016855584.1 8 - 9 1125 76
XM_017000096.1 XP_016855585.1 8 - 9 1135 76
GenBank mRNA AF086922.1 - 7 - 8 918 76
AF100760.1 - 7 - 8 918 76
AF125465.1 - 7 - 8 926 76
AF130736.1 - 7 - 8 869 76
AL136725.1 - 7 - 8 931 76
BC002399.2 - 7 - 8 867 76
BC003132.1 - 7 - 8 875 76
BC006147.1 - 7 - 8 901 76
BC007280.1 - 7 - 8 944 76
BC015153.1 - 7 - 8 943 76
BC021139.1 - 7 - 8 916 76

View Full Product Details