Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL10
Entrez Gene ID
317719
Gene Name
kelch like family member 10
Gene Aliases
SPGF11
Chromosome Location
Chr.17: 41835685 - 41848347 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152467.3 NP_689680.2 3 - 4 1446 70
XM_011524706.2 XP_011523008.1 5 - 6 1579 70
GenBank mRNA AK057224.1 - 3 - 4 1409 70
AK301797.1 - 3 - 4 1350 70
AY495339.1 - 3 - 4 1428 70
BC067753.1 - 3 - 4 1433 70

View Full Product Details