The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014008.4 | NP_054727.1 | 2 - 3 | 392 | 63 |
XM_005272599.3 | XP_005272656.1 | 2 - 3 | 324 | 63 | |
GenBank mRNA | AJ005890.1 | - | 2 - 3 | 308 | 63 |
AK291976.1 | - | 2 - 3 | 353 | 63 | |
AK296911.1 | - | 2 - 3 | 363 | 63 | |
AK298264.1 | - | 2 - 3 | 387 | 63 | |
AK316375.1 | - | 2 - 3 | 338 | 63 | |
BC000972.2 | - | 2 - 3 | 339 | 63 | |
BC011675.2 | - | 2 - 3 | 223 | 63 | |
BG912845.1 | - | 2 - 3 | 378 | 63 | |
DC340647.1 | - | 2 - 3 | 389 | 63 |