Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP6R1
Entrez Gene ID
22870
Gene Name
protein phosphatase 6 regulatory subunit 1
Gene Aliases
KIAA1115, PP6R1, SAP190, SAPS1
Chromosome Location
Chr.19: 55229779 - 55258670 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014931.3 NP_055746.3 12 - 13 1723 142
GenBank mRNA AB029038.2 - 12 - 13 2048 142
BC002799.2 - 11 - 12 1325 142
BC068014.1 - 11 - 12 1387 142
BC094753.1 - 12 - 13 1585 142

View Full Product Details