The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014931.3 | NP_055746.3 | 18 - 19 | 2368 | 81 |
GenBank mRNA | AB029038.2 | - | 18 - 19 | 2693 | 81 |
AK126810.1 | - | 1 - 2 | 1120 | 81 | |
BC002799.2 | - | 17 - 18 | 1970 | 81 | |
BC007629.1 | - | 2 - 3 | 214 | 81 | |
BC068014.1 | - | 17 - 18 | 2032 | 81 | |
BC094753.1 | - | 18 - 19 | 2230 | 81 |